N-Acetyl-L-cysteine
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
003631-25G
003631-100G
FLUO003631-25GEA
0
GBP
FLUO003631-25G
FLUO003631-100G
N-Acetyl-L-cysteine
N-Acetyl-L-cysteine
Formula:
C₅H₉NO₃S Boiling Pt: 371 °C (1013 hPa) Melting Pt: 106…108 °C Density: 1.48 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00004880 CAS Number: 616-91-1 EINECS: 210-498-3 Merck Index: 13,00090 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...