α,α,α,α',α',α'-Hexafluoro-p-xylene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1,4-Bis(trifluoromethyl)benzene, α,α,α,β,β,β-Hexafluoro-p-xylene

003569-100G 003569-500G
FLUO003569-100GEA 0 GBP
FLUO003569-100G FLUO003569-500G
α,α,α,α',α',α'-Hexafluoro-p-xylene
α,α,α,α',α',α'-Hexafluoro-p-xylene
Formula: C₈H₄F₆
Boiling Pt: 113…115 °C (1013 hPa)
Melting Pt: 3 °C
Density: 1,3808 g/cm³ (25 °C)
MDL Number: MFCD00000402
CAS Number: 433-19-2
EINECS: 207-086-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR