1H,1H,2H,2H-Perfluoro-1-iodooctane

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

003300-25G 003300-100G 003300-250G 003300-5G
FLUO003300-25GEA 0 GBP
FLUO003300-25G FLUO003300-100G FLUO003300-250G FLUO003300-5G
1H,1H,2H,2H-Perfluoro-1-iodooctane
1H,1H,2H,2H-Perfluoro-1-iodooctane
Formula: C₈H₄F₁₃I
Boiling Pt: 179 °C (1013 hPa)
Melting Pt: 20,5 °C
Density: 1,91 g/cm³ (25 °C)
MDL Number: MFCD00039410
CAS Number: 2043-57-4
EINECS: 218-056-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR