α,α,α-Trifluoro-3,5-diaminotoluene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 5-(Trifluoromethyl)-1,3-phenylenediamine, 5-(Trifluoromethyl)-m-phenylenediamine, 5-Trifluoromethyl-1,3-phenylenediamine, 5-(Trifluoromethyl)benzene-1,3-diamine, 3,5-Diaminobenzotrifluoride

003180-5G 003180-1G 003180-25G
FLUO003180-5GEA 0 GBP
FLUO003180-5G FLUO003180-1G FLUO003180-25G
α,α,α-Trifluoro-3,5-diaminotoluene
α,α,α-Trifluoro-3,5-diaminotoluene
Formula: C₇H₇F₃N₂
MDL Number: MFCD00014776
CAS Number: 368-53-6
EINECS: 206-708-8
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR