α,α,α-Trifluoro-3,4-diaminotoluene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4-(Trifluoromethyl)-1,2-phenylenediamine, 4-(Trifluoromethyl)benzene-1,2-diamine, 3,4-Diaminobenzotrifluoride, 4-(Trifluoromethyl)-o-phenylenediamine

003179-5G 003179-25G
FLUO003179-5GEA 0 GBP
FLUO003179-5G FLUO003179-25G
α,α,α-Trifluoro-3,4-diaminotoluene
α,α,α-Trifluoro-3,4-diaminotoluene
Formula: C₇H₇F₃N₂
MW: 176.14
Boiling Pt: 101…102 °C (7 mmHg)
Melting Pt: 58
MDL Number: MFCD00042456
CAS Number: 368-71-8
UN: 2811
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR