4-(Methylthio)aniline
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
4-(Methylmercapto)aniline
003091-25G
003091-100G
FLUO003091-25GEA
0
GBP
FLUO003091-25G
FLUO003091-100G
4-(Methylthio)aniline
4-(Methylthio)aniline
Formula:
C₇H₉NS MW: 139.22 g/mol Boiling Pt: 74 °C (0.1 mmHg) Melting Pt: 156…158 °C Density: 1.1379 g/cm³ (20 °C) |
MDL Number:
MFCD00007889 CAS Number: 104-96-1 EINECS: 203-256-3 UN: 2810 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...