Tetrafluorophthalic acid
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
003055.5G
003055-25G
003055-10G
003055-100G
FLUO003055.5GEA
18.8
GBP
FLUO003055.5G
FLUO003055-25G
FLUO003055-10G
FLUO003055-100G
Tetrafluorophthalic acid
Tetrafluorophthalic acid
Formula:
C₈H₂F₄O₄ MW: 238.1 g/mol Boiling Pt: 341 °C (1013 hPa) Melting Pt: 155…156 °C |
MDL Number:
MFCD00002407 CAS Number: 652-03-9 EINECS: 211-483-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...