Furfuryl mercaptan

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

002796-25G 002796-100G
FLUO002796-25GEA 0 GBP
FLUO002796-25G FLUO002796-100G
Furfuryl mercaptan
Furfuryl mercaptan
Formula: C₅H₆OS
Boiling Pt: 155 °C (1013 hPa)
Density: 1,1339 g/cm³ (20 °C)
Flash Pt: 51 °C
MDL Number: MFCD00003254
CAS Number: 98-02-2
EINECS: 202-628-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR