Thiazole

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

002766-5G 002766-25G
FLUO002766-5GEA 0 GBP
FLUO002766-5G FLUO002766-25G
Thiazole
Thiazole
Formula: C₃H₃NS
Boiling Pt: 116.8 °C (1013 hPa)
Melting Pt: –34 °C
Density: 1.2 g/cm³ (20 °C)
MDL Number: MFCD00005315
CAS Number: 288-47-1
EINECS: 206-021-3
Merck Index: 12,09442

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR