4-Methylthiazole

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

002765-25G 002765-100G
FLUO002765-25GEA 0 GBP
FLUO002765-25G FLUO002765-100G
4-Methylthiazole
4-Methylthiazole
Formula: C₄H₅NS
MW: 99.16 g/mol
Boiling Pt: 133 °C (1013 hPa)
Density: 1,1187 g/cm³ (20 °C)
MDL Number: MFCD00005340
CAS Number: 693-95-8
EINECS: 211-764-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR