4,5-Dimethylthiazole
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
002761-25G
002761-5G
FLUO002761-25GEA
0
GBP
FLUO002761-25G
FLUO002761-5G
4,5-Dimethylthiazole
4,5-Dimethylthiazole
Formula:
C₅H₇NS MW: 113.18 g/mol Boiling Pt: 149…153 °C (1013 hPa) Density: 1,0828 g/cm³ (20 °C) |
MDL Number:
MFCD00005336 CAS Number: 3581-91-7 EINECS: 222-703-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...