2,4-Dimethylthiazole
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
002760-25G
002760-100G
002760-5G
FLUO002760-25GEA
0
GBP
FLUO002760-25G
FLUO002760-100G
FLUO002760-5G
2,4-Dimethylthiazole
2,4-Dimethylthiazole
Formula:
C₅H₇NS MW: 113.18 g/mol Boiling Pt: 144 °C (1013 hPa) Density: 1,0601 g/cm³ (15 °C) |
MDL Number:
MFCD00014509 CAS Number: 541-58-2 EINECS: 208-786-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...