4,4'-Difluorobenzophenone

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: Bis(4-Fluorophenyl)methanone, Bis(4-fluorophenyl) ketone

002692-50G 002692-250G 002692-1KG
FLUO002692-50GEA 0 GBP
FLUO002692-50G FLUO002692-250G FLUO002692-1KG
4,4'-Difluorobenzophenone
4,4'-Difluorobenzophenone
Formula: C₁₃H₈F₂O
MW: 218.2 g/mol
Boiling Pt: 170…172 °C (10 mmHg)
Melting Pt: 140…141 °C
Density: 1.39 g/cm³ (20 °C)
MDL Number: MFCD00000353
CAS Number: 345-92-6
EINECS: 206-466-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR