4,4'-Difluorobenzhydrol

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4,4'-Difluorophenyl phenyl carbinol, 4,4'-Difluorobenzhydryl alcohol, 4,4'-Difluoro-α-phenylbenzyl alcohol, Bis(4-fluorophenyl)carbinol, 4,4'-Difluorodiphenylmethanol, Bis(4-fluorophenyl)methanol

002691-25G 002691-100G
FLUO002691-25GEA 0 GBP
FLUO002691-25G FLUO002691-100G
4,4'-Difluorobenzhydrol
4,4'-Difluorobenzhydrol
Formula: C₁₃H₁₀F₂O
MW: 220.22 g/mol
Boiling Pt: 305
Melting Pt: 47
Density: 1 g/cm³ (20 °C)
MDL Number: MFCD00000357
CAS Number: 365-24-2
EINECS: 206-671-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR