4-Ethoxyphenyl isothiocyanate

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1-Ethoxy-4-isothiocyanato-benzene, 4-Ethoxyphenylisothiocyanate

002562-5G 002562-25G
FLUO002562-5GEA 0 GBP
FLUO002562-5G FLUO002562-25G
4-Ethoxyphenyl isothiocyanate
4-Ethoxyphenyl isothiocyanate
Formula: C₉H₉NOS
MW: 179.24 g/mol
Boiling Pt: 146 °C (19 mmHg)
Melting Pt: 75…76 °C
MDL Number: MFCD00041096
CAS Number: 3460-49-9
EINECS: 222-410-0
UN: 2923
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR