3-Chloropropyl isothiocyanate

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

002535-1G 002535-5G
FLUO002535-1GEA 0 GBP
FLUO002535-1G FLUO002535-5G
3-Chloropropyl isothiocyanate
3-Chloropropyl isothiocyanate
Formula: C₄H₆ClNS
MDL Number: MFCD00041131
CAS Number: 2799-72-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR