4-Chlorobenzyl isothiocyanate

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1-Chloro-4-(isothiocyanatomethyl)benzene

002529-5G 002529-1G
FLUO002529-5GEA 0 GBP
FLUO002529-5G FLUO002529-1G
4-Chlorobenzyl isothiocyanate
4-Chlorobenzyl isothiocyanate
Formula: C₈H₆ClNS
Boiling Pt: 105…108 °C (0.3 mmHg)
Density: 1,291 g/cm³ (20 °C)
MDL Number: MFCD00041116
CAS Number: 3694-45-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR