3-Chlorobenzyl isothiocyanate
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
002528-5G
002528-1G
FLUO002528-5GEA
0
GBP
FLUO002528-5G
FLUO002528-1G
3-Chlorobenzyl isothiocyanate
3-Chlorobenzyl isothiocyanate
Formula:
C₈H₆ClNS |
MDL Number:
MFCD00046820 CAS Number: 3694-58-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...