(±)-1-Phenyl-2,2,2-trifluoroethanol

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: α-(Trifluoromethyl)benzyl alcohol, 2,2,2-Trifluoro-1-phenylethan-1-ol

002360-5G 002360-25G
FLUO002360-5GEA 0 GBP
FLUO002360-5G FLUO002360-25G
(±)-1-Phenyl-2,2,2-trifluoroethanol
(±)-1-Phenyl-2,2,2-trifluoroethanol
Formula: C₆H₅CH(OH)CF₃
MW: 176.14 g/mol
MDL Number: MFCD00004498
CAS Number: 340-05-6
EINECS: 206-429-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR