3-Fluorophthalic anhydride

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

002357-5G 002357-25G 002357-100G
FLUO002357-5GEA 0 GBP
FLUO002357-5G FLUO002357-25G FLUO002357-100G
3-Fluorophthalic anhydride
3-Fluorophthalic anhydride
Formula: C₈H₃FO₃
Boiling Pt: 351 °C (1013 hPa)
Melting Pt: 162…163 °C
MDL Number: MFCD00039696
CAS Number: 652-39-1
EINECS: 211-491-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR