3-Fluorophthalic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

002356-5G 002356-25G
FLUO002356-5GEA 0 GBP
FLUO002356-5G FLUO002356-25G
3-Fluorophthalic acid
3-Fluorophthalic acid
Formula: C₈H₅FO₄
MW: 184.12 g/mol
MDL Number: MFCD00039524
CAS Number: 1583-67-1
EINECS: 216-433-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR