5-Fluoro-2-nitrobenzotrifluoride
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
5-Fluoro-2-nitrobenzotrifluoride, α,α,α,5-Tetrafluoro-2-nitrotoluene, 1-Fluoro-4-nitro-3-(trifluoromethyl)benzene
002250-100G
002250-25G
FLUO002250-100GEA
0
GBP
FLUO002250-100G
FLUO002250-25G
5-Fluoro-2-nitrobenzotrifluoride
5-Fluoro-2-nitrobenzotrifluoride
Formula:
C₇H₃F₄NO₂ MW: 209.1 g/mol Boiling Pt: 198…199 °C (1013 hPa) Melting Pt: 23.5 °C Density: 1.49 g/cm³ (20 °C) |
MDL Number:
MFCD00039741 CAS Number: 393-09-9 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...