3,5-Bis(trifluoromethyl)benzyl bromide

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

002102-5G 002102-25G 002102-100G
FLUO002102-5GEA 0 GBP
FLUO002102-5G FLUO002102-25G FLUO002102-100G
3,5-Bis(trifluoromethyl)benzyl bromide
3,5-Bis(trifluoromethyl)benzyl bromide
Formula: C₉H₅BrF₆
Boiling Pt: 136…140 °C (14 mmHg)
Density: 1.67 g/cm³ (20 °C)
MDL Number: MFCD00009905
CAS Number: 32247-96-4
EINECS: 250-971-1
UN: 3265
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR