3,5-Bis(trifluoromethyl)benzaldehyde
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
002097-25G
002097-100G
002097-10G
FLUO002097-25GEA
0
GBP
FLUO002097-25G
FLUO002097-100G
FLUO002097-10G
3,5-Bis(trifluoromethyl)benzaldehyde
3,5-Bis(trifluoromethyl)benzaldehyde
Formula:
C₉H₄F₆O Boiling Pt: 47…48 °C (0,2 mmHg) Melting Pt: –2 °C Density: 1,46 g/cm³ (20 °C) |
MDL Number:
MFCD00010206 CAS Number: 401-95-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...