α,α,α-Trifluoro-o-toluonitrile
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
001973-25G
001973-100G
001973-250G
FLUO001973-25GEA
0
GBP
FLUO001973-25G
FLUO001973-100G
FLUO001973-250G
α,α,α-Trifluoro-o-toluonitrile
α,α,α-Trifluoro-o-toluonitrile
Formula:
C₈H₄F₃N Boiling Pt: 204 °C (1013 hPa) Melting Pt: 18 °C Density: 1.2886 g/cm³ (20 °C) |
MDL Number:
MFCD00001791 CAS Number: 447-60-9 EINECS: 207-184-3 UN: 3276 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...