3,3,3-Trifluoropropionic acid
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
3,3,3-Trifluoropropanoic acid
001558-5G
001558-25G
001558-100G
FLUO001558-5GEA
0
GBP
FLUO001558-5G
FLUO001558-25G
FLUO001558-100G
3,3,3-Trifluoropropionic acid
3,3,3-Trifluoropropionic acid
Formula:
C₃H₃F₃O₂ MW: 128.05 g/mol Boiling Pt: 153 Melting Pt: 0 Density: 1.441 g/cm³ (27 °C) |
MDL Number:
MFCD00153292 CAS Number: 2516-99-6 UN: 3265 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...