α,α,α-Trifluoro-m-toluonitrile
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
3-Cyanobenzotrifluoride, m-(Trifluoromethyl)benzonitrile, 3-(Trifluoromethyl)benzonitrile
001425-500G
001425-100G
FLUO001425-500GEA
0
GBP
FLUO001425-500G
FLUO001425-100G
α,α,α-Trifluoro-m-toluonitrile
α,α,α-Trifluoro-m-toluonitrile
Formula:
C₈H₄F₃N MW: 171.12 g/mol Boiling Pt: 189 °C (1013 hPa) Melting Pt: 19…22 °C Density: 1.2821 g/cm³ (20 °C) |
MDL Number:
MFCD00001807 CAS Number: 368-77-4 EINECS: 206-711-4 UN: 3276 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...