α,α,α-Trifluoro-p-toluonitrile

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4-(Trifluoromethyl)benzonitrile, 4-Cyanobenzotrifluoride, p-(Trifluoromethyl)benzonitrile

001411-25G 001411-100G 001411-250G
FLUO001411-25GEA 0 GBP
FLUO001411-25G FLUO001411-100G FLUO001411-250G
α,α,α-Trifluoro-p-toluonitrile
α,α,α-Trifluoro-p-toluonitrile
Formula: C₈H₄F₃N
MW: 171.12 g/mol
Boiling Pt: 183…185 °C (1013 hPa)
Melting Pt: 35…38 °C
Density: 1.2784 g/cm³ (20 °C)
Flash Pt: 72 °C (closed cup)
MDL Number: MFCD00001826
CAS Number: 455-18-5
EINECS: 207-239-1
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR