o-Tolylacetic acid
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
2-Tolylacetic acid, (2-Methylphenyl)acetic acid
001393-25G
001393-100G
FLUO001393-25GEA
0
GBP
FLUO001393-25G
FLUO001393-100G
o-Tolylacetic acid
o-Tolylacetic acid
Formula:
(C₆H₄CH₃)CH₂COOH MW: 150.18 g/mol Boiling Pt: 283 Melting Pt: 86…87 °C |
MDL Number:
MFCD00004328 CAS Number: 644-36-0 EINECS: 211-416-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...