m-Tolylacetic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-(m-Tolyl)acetic acid, (3-Methylphenyl)acetic acid

001325-25G 001325-100G
FLUO001325-25GEA 0 GBP
FLUO001325-25G FLUO001325-100G
m-Tolylacetic acid
m-Tolylacetic acid
Formula: (C₆H₄(CH₃))CH₂COOH
MW: 150.18 g/mol
Boiling Pt: 120…123 °C (26 mmHg)
Melting Pt: 64
MDL Number: MFCD00004340
CAS Number: 621-36-3
EINECS: 210-683-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR