2-Acetyl-5-bromothiophene
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
1-(5-Bromo-2-thienyl)-1-ethanone
001260-25G
001260-100G
FLUO001260-25GEA
0
GBP
FLUO001260-25G
FLUO001260-100G
2-Acetyl-5-bromothiophene
2-Acetyl-5-bromothiophene
Formula:
C₆H₅BrOS MW: 205.08 g/mol Boiling Pt: 253 Melting Pt: 92…93 °C |
MDL Number:
MFCD00014528 CAS Number: 5370-25-2 EINECS: 226-363-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...