3,3,3-Triphenylpropionic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3,3,3-Triphenylpropanoic acid, 3,3,3-Triphenylpropanoic acid

001032-5G 001032-25G
FLUO001032-5GEA 0 GBP
FLUO001032-5G FLUO001032-25G
3,3,3-Triphenylpropionic acid
3,3,3-Triphenylpropionic acid
Formula: C₂₁H₁₈O₂
MW: 302.37 g/mol
Melting Pt: 184
Density: 1.264 g/cm³ (20 °C)
MDL Number: MFCD00002713
CAS Number: 900-91-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR