3,5-Bis(trifluoromethyl)phenol
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
001013-25G
001013-10G
001013-50G
FLUO001013-25GEA
0
GBP
FLUO001013-25G
FLUO001013-10G
FLUO001013-50G
3,5-Bis(trifluoromethyl)phenol
3,5-Bis(trifluoromethyl)phenol
Formula:
C₈H₄F₆O Boiling Pt: 176 °C (1013 hPa) Melting Pt: 21 °C Density: 1,51 g/cm³ (20 °C) |
MDL Number:
MFCD00000386 CAS Number: 349-58-6 EINECS: 206-488-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...