2-(Trifluoromethyl)pyridine ≥99%
Supplier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
L19554.03
L19554.MD
L19554.03EA
65.4
GBP
L19554.03
L19554.MD
2-(Trifluoromethyl)pyridine ≥99%
2-(Trifluoromethyl)pyridine
Purity: 99%
CAS No.: 368-48-9
Molecular Formula: C6H4F3N
Formula Weight: 147.10
Flash Point: 42°(108°F)
Refractive Index: 1.4180
Boiling Point: 139-141°
Beilstein Reference: 3112
UN No.: 1993
MDL No.: MFCD02183562
Formula:
C₆H₄F₃N MW: 147.1 g/mol Boiling Pt: 139…141 °C Density: 1.28 Flash Pt: 42 °C (108 °F) Storage Temperature: Ambient |
MDL Number:
MFCD02183562 CAS Number: 368-48-9 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...