2,3-Bis(trifluoromethyl)pyridine ≥97%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

L19445.03 L19445.MD
L19445.03EA 57.8 GBP
L19445.03 L19445.MD
2,3-Bis(trifluoromethyl)pyridine ≥97%
2,3-Bis(trifluoromethyl)pyridine

Purity: 97%
CAS No.: 1644-68-4
Molecular Formula: C7H3F6N
Formula Weight: 215.10
Flash Point: 70°(158°F)
Refractive Index: 1.3940
Melting Point: -2°
Boiling Point: 85°/18mm
Beilstein Reference: 1622028
UN No.: 2810
MDL No.: MFCD03093763

Formula: C₇H₃F₆N
Boiling Pt: 158 °C (1013 hPa)
Melting Pt: –2 °C
MDL Number: MFCD03093763
CAS Number: 1644-68-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR