Indole-4-carbaldehyd ≥97%
Supplier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
H28019.06
H28019.03
H28019.06EA
582
GBP
H28019.06
H28019.03
Indole-4-carbaldehyd ≥97%
Indole-4-carbaldehyd
Purity: 97%
CAS No.: 1074-86-8
Molecular Formula: C9H7NO
Formula Weight: 145.16
Melting Point: 143-144°
MDL No.: MFCD01632221
Formula:
C₉H₇NO Melting Pt: 143…144 °C Storage Temperature: Ambient |
MDL Number:
MFCD01632221 CAS Number: 1074-86-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...