4-(Trifluoromethyl)anilinium chloride ≥97%

Supplier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: 4-Aminobenzotrifluoride HCl, 4-Aminobenzotrifluoride hydrochloride, α,α,α-Trifluoro-p-toluidine HCl, α,α,α-Trifluoro-p-toluidine hydrochloride, 4-(Trifluoromethyl)aniline hydrochloride

A18329.06 A18329.14
A18329.06EA 31.2 GBP
A18329.06 A18329.14
4-(Trifluoromethyl)anilinium chloride ≥97%
4-(Trifluoromethyl)anilinium chloride

Purity: 97% CAS No.: 90774-69-9 Molecular Formula: C7H6F3N.HCl Formula Weight: 197.59 Melting Point: ca 240° dec. Beilstein Reference: 3629991 UN No.: 2811 MDL No.: MFCD00007897

Formula: C₇H₇ClF₃N
MW: 197.59 g/mol
Melting Pt: ∼ 240 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00007897
CAS Number: 90774-69-9
EINECS: 292-679-7
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR