4-Phenoxyaniline ≥97%

Supplier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: 1-Amino-4-phenoxybenzol

A16048.22 A16048.14 A16048.36
A16048.22EA 52.6 GBP
A16048.22 A16048.14 A16048.36
4-Phenoxyaniline ≥97%
4-Phenoxyaniline

Purity: 97% CAS No.: 139-59-3 Molecular Formula: C12H11NO Formula Weight: 185.23 Melting Point: 82-86° Boiling Point: 140°/2mm UN No.: 3077 MDL No.: MFCD00007862

Formula: C₁₂H₁₁NO
MW: 185.23 g/mol
Boiling Pt: 140 °C (2 mmHg)
Melting Pt: 82…86 °C
Storage Temperature: Ambient
MDL Number: MFCD00007862
CAS Number: 139-59-3
EINECS: 205-367-2
ADR: 9,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR