Tris(4-methoxyphenyl)phosphine ≥98%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: Tris(p-methoxyphenyl)phosphine

A15596.03 A15596.06 A15596.14
A15596.03EA 26.6 GBP
A15596.03 A15596.06 A15596.14
Tris(4-methoxyphenyl)phosphine ≥98%
Tris(4-methoxyphenyl)phosphine

Purity: 98% CAS No.: 855-38-9 Molecular Formula: C21H21O3P Formula Weight: 352.37 Melting Point: 130-132° Beilstein Reference: 2815911

Formula: C₂₁H₂₁O₃P
Boiling Pt: 446 °C (1013 hPa)
Melting Pt: 130…132 °C
Storage Temperature: Ambient
MDL Number: MFCD00014896
CAS Number: 855-38-9
EINECS: 212-723-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR