4-Nitrophthalimide ≥98%
Supplier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
A12040.06
A12040.14
A12040.22
A12040.06EA
25.9
GBP
A12040.06
A12040.14
A12040.22
4-Nitrophthalimide ≥98%
4-Nitrophthalimide
Purity: 98%
CAS No.: 89-40-7
Molecular Formula: C8H4N2O4
Formula Weight: 192.13
Melting Point: 200-204°
Beilstein Reference: 180224
Formula:
C₈H₄N₂O₄ Melting Pt: 200…204 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005884 CAS Number: 89-40-7 EINECS: 201-905-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...