1-(4-Methylphenyl)-2-thiourea ≥97%

Supplier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: 4-Methylphenylthiourea, N-(p-Tolyl)thiourea, N-(4-Methylphenyl)thiourea, p-Tolylthiourea, 1-(p-Tolyl)thiourea, 1-(4-Tolyl)thiourea

A11098.03 A11098.06
A11098.03EA 22.3 GBP
A11098.03 A11098.06
1-(4-Methylphenyl)-2-thiourea ≥97%
1-(4-Methylphenyl)-2-thiourea

Purity: 97% CAS No.: 622-52-6 Molecular Formula: C8H10N2S Formula Weight: 166.25 Odour: Odourless Melting Point: 182-186° Beilstein Reference: 2086619 UN No.: 2811 MDL No.: MFCD00041190

Formula: (C₆H₄(CH₃))NC(S)NH₂
MW: 166.25 g/mol
Boiling Pt: 295 °C (1013 hPa)
Melting Pt: 182…186 °C
Storage Temperature: Ambient
MDL Number: MFCD00041190
CAS Number: 622-52-6
EINECS: 210-740-8
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR