4-Chlorobutyronitrile 97%

Supplier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

329940050 329941000 329945000
ACRO329940050EA 16.6 GBP
ACRO329940050 ACRO329941000 ACRO329945000
4-Chlorobutyronitrile 97%
4-Chlorobutyronitrile
Formula: C₄H₆ClN
MW: 103.55 g/mol
Boiling Pt: 195…197 °C (760 mmHg)
Melting Pt: -44 °C
Density: 1.08 g/cm³
Flash Pt: 85 °C
Storage Temperature: Ambient
MDL Number: MFCD00001972
CAS Number: 628-20-6
EINECS: 211-031-6
UN: 3276
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4420 to 1.4440 (20°C, 589 nm)
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR