L(-)-Proline, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5


M-Clarity™ quality level = MQ200

P5607-100G P5607-1KG P5607-25G
SIALP5607-100GEA 398 GBP
SIALP5607-100G SIALP5607-1KG SIALP5607-25G
L(-)-Proline, Sigma-Aldrich®
L(-)-Proline
Formula: C₅H₉NO₂
Boiling Pt: 245 °C (1013 hPa)
Melting Pt: 220…222 °C
Density: 1.35 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064318
CAS Number: 147-85-3
EINECS: 205-702-2
Merck Index: 13,07871

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR