4-Fluoro-α-methylbenzyl alcohol

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: p-Fluoro-α-methylbenzyl alcohol, 1-(4-Fluorophenyl)ethyl Alcohol, 1-(4-Fluorophenyl)ethanol, 4-Fluorophenyl methyl carbinol

001295-5G 001295-25G
FLUO001295-5GEA 0 GBP
FLUO001295-5G FLUO001295-25G
4-Fluoro-α-methylbenzyl alcohol
4-Fluoro-α-methylbenzyl alcohol
Formula: C₈H₉FO
MW: 140.16 g/mol
Boiling Pt: 49.5 °C (0.4 mmHg)
Melting Pt: 9.3
Density: 1.1206 g/cm³ (20 °C)
MDL Number: MFCD00004515
CAS Number: 403-41-8
EINECS: 206-959-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR