3-Bromo-α-methylbenzyl alcohol

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3-Bromophenyl methyl carbinol, m-Bromo-α-methylbenzyl alcohol, 1-(3-Bromophenyl)ethanol

003550-5G 003550-1G 003550-25G
FLUO003550-5GEA 0 GBP
FLUO003550-5G FLUO003550-1G FLUO003550-25G
3-Bromo-α-methylbenzyl alcohol
3-Bromo-α-methylbenzyl alcohol
Formula: C₈H₉BrO
MW: 201.06 g/mol
MDL Number: MFCD00021863
CAS Number: 52780-14-0
EINECS: 258-177-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR