(±)-4-Fluoromandelic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4-Fluoromandelic Acid, (±)-4-Fluorophenylglycolic acid

006259-5G 006259-25G
FLUO006259-5GEA 0 GBP
FLUO006259-5G FLUO006259-25G
(±)-4-Fluoromandelic acid
(±)-4-Fluoromandelic acid
Formula: C₈H₇FO₃
MW: 170.14 g/mol
MDL Number: MFCD00044402
CAS Number: 395-33-5
EINECS: 206-899-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR