(±)-2-Fluoromandelic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

009726-1G 009726-5G 009726-25G
FLUO009726-1GEA 0 GBP
FLUO009726-1G FLUO009726-5G FLUO009726-25G
(±)-2-Fluoromandelic acid
(±)-2-Fluoromandelic acid
Formula: C₈H₇FO₃
MW: 170.14 g/mol
MDL Number: MFCD03095382
CAS Number: 389-31-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR