(±)-2,5-Difluoromandelic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: (±)-2,5-Difluorophenylglycolic acid

004823-1G 004823-5G
FLUO004823-1GEA 0 GBP
FLUO004823-1G FLUO004823-5G
(±)-2,5-Difluoromandelic acid
(±)-2,5-Difluoromandelic acid
Formula: C₈H₆F₂O₃
MW: 188.13 g/mol
MDL Number: MFCD00061297
CAS Number: 207853-61-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR