(±)-2-Phenylpropanal, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
DL-2-Phenylpropionaldehyde, (±)-2-Phenylpropionaldehyde, (±)-Hydratropaldehyde
M-Clarity™ quality level = MQ300
W288608-10KG-K
W288608-1KG-K
W288608-5KG-K
SIALW288608-10KG-KEA
1520
GBP
SIALW288608-10KG-K
SIALW288608-1KG-K
SIALW288608-5KG-K
(±)-2-Phenylpropanal, Sigma-Aldrich®
(±)-2-Phenylpropanal
Formula:
C₉H₁₀O MW: 134.18 g/mol Density: 1.01 g/cm³ (20 °C) |
MDL Number:
MFCD00006973 CAS Number: 93-53-8 EINECS: 202-255-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...