BioHazard Spill Trays
Supplier: CLEAVER SCIENTIFIC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CSR-TO1 CSR-TO5 CSR-TO6
CLEVCSR-TO1EA 110 GBP
CLEVCSR-TO1 CLEVCSR-TO5 CLEVCSR-TO6
BioHazard Spill Trays
Trays/Dishes/Pots Spillage Trays/Pans
Spilltrays provide a re-usable work area with the added benefit of safe containment of spillages and lab bench protection. All trays supplied with one free liner when purchased.
  • One free liner included
  • White with biohazard symbol
Additional environmentally friendly APET liners are provided in packs of 25.